Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tesaglitazar
GREENER_CHOICE

Catalog No. 396510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-651-0 10 mg
39-655-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-651-0 Supplier Tocris Bioscience™ Supplier No. 3965/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

PPARα/γ agonist

Dual-specificity PPARα/γ agonist (IC50 values are 0.35 and 3.8 μM for PPARγ and PPARα respectively). Prevents atherosclerosis progression in E3L.CETP transgenic mice. Also reduces insulin resistance in obese Zucker rats. Orally active.

Chemical Identifiers

CAS 251565-85-2
Molecular Formula C20H24O7S

Specifications

Quantity 10 mg
Formula Weight 408.47
Purity >98%
Molecular Weight (g/mol) 408.47
Chemical Name or Material Tesaglitazar
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.