Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TESPA1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB190198 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB190198 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB190198 Supplier Novus Biologicals Supplier No. NBP281842JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TESPA1 Polyclonal antibody specifically detects TESPA1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen TESPA1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias hypothetical protein LOC9840, KIAA0748
Host Species Rabbit
Immunogen TESPA1 antibody was raised against an 18 amino acid peptide near the amino terminus of human TESPA1. The immunogen is located within amino acids 20 - 70 of TESPA1. Amino Acid Squence: LQDVPDPEPSSLDDVFQE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 9840
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.