Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Testosterone Antibody (3T16), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309569 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309569 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309569 Supplier Novus Biologicals Supplier No. NBP250332MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Testosterone Monoclonal antibody specifically detects Testosterone in Nonspecies specific samples. It is validated for ELISA, Radioimmune Assays

Specifications

Antigen Testosterone
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 3T16
Conjugate mFluor Violet 450 SE
Dilution ELISA, Radioimmunoassay
Formulation 50mM Sodium Borate
Gene Alias 17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone
Host Species Mouse
Immunogen Testosterone-3-(O-carboxymethyl)oxime coupled to bovine serum albumin
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Developmental Biology
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.