Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Testosterone Antibody (BGN/06/902), Alexa Fluor™ 532
SDP

Catalog No. N1062942A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1062942A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1062942A32 Supplier Novus Biologicals Supplier No. NB10062942AF532

Mouse Monoclonal Antibody

Testosterone Monoclonal specifically detects Testosterone in Non-species specific samples. It is validated for ELISA.

Specifications

Antigen Testosterone
Applications ELISA
Classification Monoclonal
Clone BGN/06/902
Conjugate Alexa Fluor 532
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone
Host Species Mouse
Immunogen Testosterone-11-HS-BSA
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cellular Signaling, Developmental Biology
Primary or Secondary Primary
Test Specificity This product is specific for Testosterone. It has shown the following reactivities. Testosterone 100% 19 Nortestosterone 15% Dihydrotestosterone <1% Testosterone promotes the development and maintenance of male secondary sex characteristics and is produced primarily in the testes. It is also produced in small amounts by the adrenal glands, ovaries and placenta. Synthetic derivatives of testosterone are used therapeutically in the treatment of breast cancer and for hormone replacement therapy.
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.