Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Testosterone Antibody (BGN/3G8A4), FITC, Novus Biologicals™
SDP

Catalog No. NB10062943F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062943F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062943F Supplier Novus Biologicals Supplier No. NB10062943F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Testosterone Monoclonal specifically detects Testosterone in Human, Non-species specific samples. It is validated for ELISA, Flow Cytometry.

Specifications

Antigen Testosterone
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone BGN/3G8A4
Conjugate FITC
Dilution ELISA
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen BSA conjugated testosterone-3,-CMO
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Developmental Biology
Primary or Secondary Primary
Test Specificity This product recognises testosterone, a steroid hormone and most potent of the naturally occurring androgens. Testosterone promotes the development and maintenance of male secondary sex characteristics and is produced primarily in the testes, but is also produced in small amounts by the adrenal glands, ovaries and placenta. Synthetic derivatives of testosterone are used therapeutically in the treatment of breast cancer and for hormone replacement therapy. NB100-62943 also recognises 5-a-Dihydrotestosterone (1%) and Androstenedione (1%).
Target Species Human, Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.