Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TET1 Antibody (GT1382) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294902 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294902 Supplier Novus Biologicals Supplier No. NBP325715

Mouse Monoclonal Antibody

TET1 Monoclonal antibody specifically detects TET1 in Human samples. It is validated for Western Blot

Specifications

Antigen TET1
Applications Western Blot
Classification Monoclonal
Clone GT1382
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH7), 20% Glycerol
Gene Alias bA119F7.1, CXXC finger 6, CXXC6, CXXC6FLJ10839, FLJ41442, KIAA1676CXXC-type zinc finger protein 6, LCX, LCXEC 1.14.11.n2, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, Ten-eleven translocation 1 gene protein, ten-eleven translocation-1, tet oncogene 1
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human TET1. The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Epigenetics, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 80312
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.