Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TET3 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB259669488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259669488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259669488 Supplier Novus Biologicals Supplier No. NBP259669AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TET3 Polyclonal specifically detects TET3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TET3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 1.14.11.n2, hCG_40738, KIAA0401, methylcytosine dioxygenase TET3, MGC22014, probable methylcytosine dioxygenase TET3, tet oncogene family member 3
Gene Symbols TET3
Host Species Rabbit
Immunogen Partial recombinant protein made to an N-terminal portion of human TET3 (amino acids 1-216) [UniProt O43151]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Epigenetics, Histones and Modified Histones
Primary or Secondary Primary
Gene ID (Entrez) 200424
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.