Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TET3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220602 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220602 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220602 Supplier Novus Biologicals Supplier No. NBP220602
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TET3 Polyclonal antibody specifically detects TET3 in Human, Mouse, Rat, Bovine, Xenopus samples. It is validated for Western Blot, ChIP assay, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry Whole-Mount.

Specifications

Antigen TET3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Flow Cytometry, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:500-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry Whole-Mount, Chromatin Immunoprecipitation (ChIP)
Formulation PBS, 20% Glycerol (pH7). with 0.025% Proclin 300
Gene Accession No. O43151
Gene Alias EC 1.14.11.n2, hCG_40738, KIAA0401, methylcytosine dioxygenase TET3, MGC22014, probable methylcytosine dioxygenase TET3, tet oncogene family member 3
Gene Symbols TET3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human TET3. The exact sequence is proprietary.
Molecular Weight of Antigen 179 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200424
Test Specificity Bovine reactivity reported in scientific literature (PMID: 24459992). Xenopus laevis reactivity reported in scientific literature (PMID: 23217707).
Target Species Human, Mouse, Rat, Bovine, Xenopus species
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.