Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetanus Toxin Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB212009 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB212009 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB212009 Supplier Novus Biologicals Supplier No. NB10063239AF750
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Tetanus Toxin Polyclonal antibody specifically detects Tetanus Toxin in Bacteria samples. It is validated for Western Blot, ELISA

Specifications

Antigen Tetanus Toxin
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias Tentoxylysin
Host Species Goat
Immunogen Purified tetanus toxin
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.