Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetanus Toxin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10062945 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062945 0.5 mL
NB017523 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10062945 Supplier Novus Biologicals Supplier No. NB10062945
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TeNT Light Chain/Tetanus Toxin Polyclonal specifically detects TeNT Light Chain/Tetanus Toxin in Bacteria samples. It is validated for ELISA.

Specifications

Antigen TeNT Light Chain/Tetanus Toxin
Applications ELISA
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation PBS with 0.09% Sodium Azide
Gene Alias Tentoxylysin, Tetanus toxin chain H, Tetanus toxin chain L, Tetanus toxin heavy chain, Tetanus toxin light chain
Gene Symbols TETX
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the amino acids 1300 - 1314 of the native molecule conjugated to Keyhole limpet Haemocyanin
Purification Method Affinity Purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1061100
Test Specificity This product recognizes the tetanus toxin, a neurotoxin that inhibits neurotransmitter release.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.