Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetanus Toxin Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $560.50
Specifications
Antigen | TeNT Light Chain/Tetanus Toxin |
---|---|
Concentration | 5.0 mg/mL |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017523
![]() |
Novus Biologicals
NB100629450.1ML |
0.1 mL |
Each for $207.00
|
|
|||||
NB10062945
![]() |
Novus Biologicals
NB10062945 |
0.5 mL |
Each for $560.50
|
|
|||||
Description
Tetanus Toxin Polyclonal specifically detects Tetanus Toxin in Bacteria samples. It is validated for ELISA.Specifications
TeNT Light Chain/Tetanus Toxin | |
ELISA 1:100-1:2000 | |
Polyclonal | |
Rabbit | |
Bacteria | |
Tentoxylysin, Tetanus toxin chain H, Tetanus toxin chain L, Tetanus toxin heavy chain, Tetanus toxin light chain | |
TETX | |
IgG | |
Affinity Purified | |
This product recognizes the tetanus toxin, a neurotoxin that inhibits neurotransmitter release. |
5.0 mg/mL | |
ELISA | |
Unconjugated | |
RUO | |
PBS with 0.09% Sodium Azide | |
1061100 | |
Synthetic peptide corresponding to the amino acids 1300 - 1314 of the native molecule conjugated to Keyhole limpet Haemocyanin | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title