Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TetR Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB600234X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600234X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600234X Supplier Novus Biologicals Supplier No. NB600234AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TetR Polyclonal specifically detects TetR in Non-species specific samples. It is validated for Western Blot, ELISA.

Specifications

Antigen TetR
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias pSC101 tetR, Tet Repressor, Tet Repressor Protein, TetR, tetracycline repressor protein
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of Escherichia coli TetR protein (between residues 25-75) [UniProt P04483]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4924774
Test Specificity Non-species specific. This antibody is used to monitor the regulation of gene expression in mammalian cells. The TetR antibody is not reactive to human or other mammalian proteins.
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.