Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TetR Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB338709 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338709 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338709 Supplier Novus Biologicals Supplier No. NB600234APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TetR Polyclonal antibody specifically detects TetR in Nonspecies specific samples. It is validated for Western Blot, ELISA

Specifications

Antigen TetR
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate APC
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias pSC101 tetR, Tet Repressor, Tet Repressor Protein, TetR, tetracycline repressor protein
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of Escherichia coli TetR protein (between residues 25-75) [UniProt P04483]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4924774
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.