Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tetrabenazine
GREENER_CHOICE

Catalog No. p-5546072
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent inhibitor of vesicular monoamine transport

Tetrabenazine is an inhibitor of vesicular monoamine transport with a 10-fold selectivity for the VMAT2 transporter over VMAT1 (IC50 values = 0.3 μM and 3.4 μM, respectively). Tetrabenazine blocks D2 receptors and dopamine uptake (IC50 = 0.12 μM) and inhibits serotonin transport by VMAT2 (IC50 = 300 nM). In an animal model of Huntington's disease, Tetrabenazine prevents the decline in motor control and reduces loss of striatal neurons. Tetrabenazine increases cFos expression in the thalamus and hippocampus, reduces brain connectivity and spontaneous locomotion and also causes behavioral depression.

Specifications

Quantity 50 mg
Product Type Tetrabenazine
Purity >98%
Molecular Weight (g/mol) 317.2
Inhibitors Tetrabenazine

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.