Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrabenazine, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
Chemical Name or Material | Tetrabenazine |
CAS | 58-46-8 |
Quantity | 1 g |
Melting Point | 124°C to 130°C |
Shipping Condition | No special requirements |
Product Line | Life science |
Molecular Formula | C19 H27 N O3 |
Physical Form | Crystalline Powder |
Color | White to Light-yellow to Beige |
Purity | ≥97.5% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction