Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabromophenolphthalein Ethyl Ester, Potassium Salt, Indicator Grade

Catalog No. p-195632
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 62637-91-6 | C22H13Br4KO4 | 700.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62637-91-6
Molecular Formula C22H13Br4KO4
Molecular Weight (g/mol) 700.06
MDL Number MFCD00011662
InChI Key WCIQBKUTYDIBJC-UHFFFAOYSA-M
PubChem CID 23689366
SMILES [K+].CCOC(=O)C1=CC=CC=C1C(C1=CC(Br)=C([O-])C(Br)=C1)=C1C=C(Br)C(=O)C(Br)=C1

Specifications

Merck Index 15, 9331
Infrared Spectrum Authentic
IUPAC Name potassium;2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)-(2-ethoxycarbonylphenyl)methyl]phenolate
Formula Weight 700.05
Grade Extra Pure
Packaging Glass bottle
Specific Extinction 660 min. (600nm, ethanol)
Color Black/Blue to Blue
Quantity 1 g
Physical Form Crystalline Powder or Granules
Chemical Name or Material Tetrabromophenolphthalein ethyl ester, potassium salt
Show More Show Less
EINECSNumber 263-661-6
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.