Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabromophthalic anhydride, 98%

Catalog No. p-7027641
Click to view available options
Quantity:
100 g
500 g

CAS: 632-79-1 | C8Br4O3 | 463.701 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-79-1
Molecular Formula C8Br4O3
Molecular Weight (g/mol) 463.701
MDL Number MFCD00005919
InChI Key QHWKHLYUUZGSCW-UHFFFAOYSA-N
PubChem CID 12443
IUPAC Name 4,5,6,7-tetrabromo-2-benzofuran-1,3-dione
SMILES C12=C(C(=C(C(=C1Br)Br)Br)Br)C(=O)OC2=O

Specifications

Melting Point 274°C to 280°C
Density 2.87
Odor Odorless
Quantity 100 g
Beilstein 18908
Sensitivity Moisture sensitive
Synonym tetrabromophthalic anhydride, bromphthal, firemaster pht 4, phthalic anhydride, tetrabromo, bromophthal, tetrabromophthalic acid anhydride, firemaster pht4, 1,3-isobenzofurandione, 4,5,6,7-tetrabromo, 4,5,6,7-tetrabromo-1,3-isobenzofurandione, pht 4
Formula Weight 463.7
Percent Purity 98%
Chemical Name or Material Tetrabromophthalic anhydride
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 211-185-4
RTECSNumber TI3350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.