Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrabromophthalic Anhydride 98%, Thermo Scientific™
Click to view available options
:
5g; Glass bottle
Specifications
Specifications
CAS | 632-79-1 |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass Bottle |
Molecular Formula | C8Br4O3 |
Quantity | 5g |
MDL Number | MFCD00005919 |
Synonym | tetrabromophthalic anhydride, bromphthal, firemaster pht 4, phthalic anhydride, tetrabromo, bromophthal, tetrabromophthalic acid anhydride, firemaster pht4, 1,3-isobenzofurandione, 4,5,6,7-tetrabromo, 4,5,6,7-tetrabromo-1,3-isobenzofurandione, pht 4 |
Solubility Information | Solubility in water: insoluble (hydrolysis to acid). Other solubilities: insoluble in ethanol,, slightly soluble in benzene,, soluble in xylene, acetic acid |
InChI Key | QHWKHLYUUZGSCW-UHFFFAOYSA-N |
Show More |
Safety and Handling
Hazard Category | MOISTURE SENSITIVE |
Hazard Statement | GHS H Statement: May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation. |
Precautionary Statement | GHS P Statement: IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 211-185-4 |
RTECSNumber | TI3350000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction