Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrachloroauric(III) acid trihydrate, ACS reagent
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Orange |
| Physical Form | Glistening Crystalline Powder or Lumps |
| Quantity | 5 g |
| Linear Formula | HAuCl4·3H2O |
| Merck Index | 15,212 |
| Synonym | Chloroauric acid trihydrate, Gold trichloride acid trihydrate |
| Solubility Information | Solubility in water: 150g/L (20°C). Other solubilities: soluble in alcohol and ether |
| Formula Weight | 393.83 |
| Percent Purity | ≥49.0% (Au) |
| Grade | ACS Reagent |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Harmful if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Con |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction