Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecane, 99%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
EcoSafPak™ Glass Bottle
Glass bottle
629-59-4
C14H30
198.39
BGHCVCJVXZWKCC-UHFFFAOYSA-N
n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
12389
CHEBI:41253
tetradecane
CCCCCCCCCCCCCC
629-59-4
C14H30
198.39
BGHCVCJVXZWKCC-UHFFFAOYSA-N
n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
12389
CHEBI:41253
tetradecane
CCCCCCCCCCCCCC
Quantity 100mL
Refractive Index 1.4280 to 1.4300
Melting Point 5.8°C
Density 0.7670g/mL
Boiling Point 252.0°C to 254.0°C
Color Colorless
Infrared Spectrum Authentic
Specific Gravity 0.767
Packaging Glass bottle
Flash Point 99°C
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)12CH3
Beilstein 01, 171
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 198.39
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Tetradecane
Show More Show Less

GHS H Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

EINECSNumber : 211-096-