Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetradecane, 99%

Catalog No. p-4510597
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
EcoSafPak™ Glass Bottle
Glass bottle

CAS: 629-59-4 | C14H30 | 198.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-59-4
InChI Key BGHCVCJVXZWKCC-UHFFFAOYSA-N
Synonym n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefine
PubChem CID 12389
ChEBI CHEBI:41253
IUPAC Name tetradecane
SMILES CCCCCCCCCCCCCC

Specifications

Melting Point 5.8°C
Color Colorless
Density 0.7670g/mL
Boiling Point 252.0°C to 254.0°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C14H30
Refractive Index 1.4280 to 1.4300
Linear Formula CH3(CH2)12CH3
Quantity 100 mL
Beilstein 01, 171
Specific Gravity 0.767
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Molecular Weight (g/mol) 198.39
Formula Weight 198.39
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetradecane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 211-096-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.