Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethoxymethane 97.0+%, TCI America™
SDP

Catalog No. T110725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T110725ML 25 mL
T11075ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T110725ML Supplier TCI America Supplier No. T110725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78-09-1
Molecular Formula C9H20O4
Molecular Weight (g/mol) 192.26
MDL Number MFCD00009221
InChI Key CWLNAJYDRSIKJS-UHFFFAOYSA-N
Synonym tetraethyl orthocarbonate, tetraethoxymethane, triethoxymethoxy ethane, tetraethylorthocarbonate, ethane, 1,1',1,1'-methanetetrayltetrakis oxy tetrakis, orthocarbonic acid, tetraethyl ester, tetraethyi orthocarbonate, tetra ethyl orthocarbonate, 1-triethoxymethoxy ethane
PubChem CID 66213
IUPAC Name (triethoxymethoxy)ethane
SMILES CCOC(OCC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 160°C
Quantity 25 mL
UN Number 3271
Formula Weight 192.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraethoxymethane
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.