Tetraethoxysilane, >99%, Alfa Aesar™

Manufacturer: Alfa Aesar 1408218

Quantity 50g

 View more versions of this product

Catalog No. AA1408218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 78-10-4
    Physical Form Liquid
    Assay Percent Range >99%
    Molecular Formula C8H20O4Si
    Formula Weight 208.33
    MDL Number MFCD00009062
    Chemical Name or Material Tetraethoxysilane

    Specifications

    Formula Weight 208.33
    Quantity 50g
    EINECS Number 201-083-8
    Physical Form Liquid
    Density 0.934g/mL
    Assay Percent Range >99%
    Molecular Formula C8H20O4Si
    MDL Number MFCD00009062
    CAS 78-10-4
    Chemical Name or Material Tetraethoxysilane

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit