Learn More
Tetraethylene glycol dimethyl ether, 98+%
Description
Tetraethylene glycol dimethyl ether is used in lithium-ion battery technology. It is associated with trifluoroethanol and used as a working pair for organic absorption heat pumps. It acts as a solvent for cleaning and degreasing due to its chemical and thermal stability. Moreover, it is suitable solvent for high temperature reactions due its high boiling point. Further, it is used in the selective adsorption of proteins during the promotion of cell adhesion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -30°C |
| Density | 1.01 g/mL |
| Boiling Point | 275°C to 276°C |
| Flash Point | 140°C (284°F) |
| Refractive Index | 1.433 |
| Quantity | 250 g |
| Merck Index | 14,9208 |
| Solubility Information | Miscible with water. |
| Formula Weight | 222.28 |
| Percent Purity | ≥98% |
| Show More |
Safety and Handling
| Hazard Category | H360Df |
| Hazard Statement | GHS H Statement H360 May damage fertility or the unborn child. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 205-594-7 |
| RTECSNumber | SB0400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.