Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrafluoroboric acid-diethyl ether complex, 50-55% w/w HBF4

Catalog No. p-7049678
Click to view available options
Quantity:
25 mL
250 mL

CAS: 67969-82-8 | C4H11BF4O | 161.935 g/mol

Tetrafluoroboric acid-diethyl ether acts as an effective catalyst used for various carbohydrate protection reactions. It is also used as a catalyst in the preparation of 4-fluoropiperidines by reaction between aldehydes and N-tosyl homoallylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67969-82-8
Molecular Formula C4H11BF4O
Molecular Weight (g/mol) 161.935
MDL Number MFCD00011345
InChI Key XFHGDBFMJCLEOW-UHFFFAOYSA-N
Synonym fluoroboric acid diethyl ether complex, tetrafluoroboric acid diethyl ether complex, hbf4.oet2, fluoroboric acid diethylether complex, boron trifluoride diethyl ether hydrofluoride
PubChem CID 11344169
IUPAC Name ethoxyethane;trifluoroborane;hydrofluoride
SMILES B(F)(F)F.CCOCC.F

Specifications

Color Colorless to Orange
Density 1.19 g/mL
Flash Point 36°C (97°F)
Linear Formula HBF4·(CH3CH2)2O
Quantity 25 mL
UN Number UN2920
Sensitivity Moisture sensitive
Merck Index 14,4149
Solubility Information Reacts in water;Miscible with dichloromethane.
Formula Weight 161.93
Concentration or Composition (by Analyte or Components) 50 to 55% w/w HBF4
Physical Form Liquid
Chemical Name or Material Tetrafluoroboric acid-diethyl ether complex
Show More Show Less
Hazard Category H226-H302-H314-H335-H336
Hazard Statement GHS H Statement
H314-H318-H226-H302-H336
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 267-983-8
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.