Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydro-3-furoic acid, 97%, Thermo Scientific™

Catalog No. AC433450050
Click to view available options
:
5g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 89364-31-8
Density 1.2140g/mL
Boiling Point 138.0°C to 140.0°C (12.0mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C5H8O3
Quantity 5g
Specific Gravity 1.214
Synonym tetrahydrofuran-3-carboxylic acid, tetrahydro-3-furoic acid, tetrahydro-furan-3-carboxylic acid, tetrahydro-3-furancarboxylic acid, tetrahydro-3-furoicacid, 3-furancarboxylic acid, tetrahydro, 3-oxolanecarboxylic acid, 3-tetrahydrofuroic acid, pubchem22077, 3-carboxytetrahydrofuran
InChI Key BOTREHHXSQGWTR-UHFFFAOYSA-N
SMILES C1COCC1C(=O)O
IUPAC Name oxolane-3-carboxylic acid
Stabilizer (250ppm BHT inhibitor)
Molecular Weight (g/mol) 116.116
PubChem CID 4661317
Formula Weight 116.12
Percent Purity 97%
Chemical Name or Material Tetrahydro-3-furoic acid
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.