Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrocannabinol Antibody (094-14066 (THC-003)), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N1062946A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1062946A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1062946A70 Supplier Novus Biologicals Supplier No. NB10062946AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tetrahydrocannabinol Monoclonal specifically detects Tetrahydrocannabinol in Non-species specific samples. It is validated for ELISA.

Specifications

Antigen Tetrahydrocannabinol
Applications ELISA
Classification Monoclonal
Clone 094-14066 (THC-003)
Conjugate Alexa Fluor 750
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen 11-Nor-delta 9-THC-acid conjugated at the carboxyl group.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity NB100-62946 reacts with Delta-9-tetrahydrocannabinol, the main psychoactive substance present in the Cannabis plant.
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.