Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tetrahydrofuran, anhydrous, 99.8+%, unstab., packaged under Argon in resealable ChemSeal™ bottles
Click to view available options
Quantity:
250 mL
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Specifications
| Melting Point | °C to 108°C |
| Appearance | Colorless liquid |
| Density | 0.889 g/cm3 at 20°C |
| Boiling Point | 66°C |
| Quantity | 1 L |
| Flash Point | -17°C (1°F) |
| Odor | Ether-like |
| Packaging | Resealable ChemSeal™ Bottle under Argon |
| Physical Form | Liquid |
| Refractive Index | 1.407 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H319-H335-H336-H351 |
| Hazard Statement | GHS H Statement H225-H351-H319-H335 |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN |
| EINECSNumber | 203-726-8 |
| RTECSNumber | LU5950000 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction