Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Low Water, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth Amber Glass Bottle with Septum

Chemical Identifiers

109-99-9
C4H8O
72.107
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
CHEBI:26911

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography, organic and biosynthesis
SMILES C1CCOC1
IUPAC Name oxolane
Formula Weight 72.11
Packaging Narrow Mouth Amber Glass Bottle with Septum
Specific Gravity 1L = 0.88kg
Flash Point −14.00°C
Quantity 1L
Chemical Name or Material Tetrahydrofuran, Low Water
Grade HPLC Grade