Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrofuran, Packaged under Argon, HPLC Grade, J.T. Baker™

Catalog No. 02004419 Shop All Avantor Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Glass Bottle

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Quantity 4 L
Flash Point −14.00°C
Packaging Narrow Mouth PVC-coated Amber Glass Bottle
Specific Gravity 1L = 0.88kg
Formula Weight 72.11
Grade HPLC
Chemical Name or Material Tetrahydrofuran, Packaged under Argon
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.