Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(acetonitrile)palladium(II) tetrafluoroborate, Pd 23.5%

Catalog No. p-7050778
Click to view available options
Quantity:
0.25 g
1 g

CAS: 21797-13-7 | C8H12B2F8N4Pd | 444.24 g/mol

Acts as a precursor for:

  • Synthesis of dendritic SCS-pincer palladium complexes.
  • Palladium complexes of click ligands.
  • Dipalladium catalysts for use in Heck cross-coupling5, Suzuki cross-coupling, and aldehyde olefination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21797-13-7
Molecular Formula C8H12B2F8N4Pd
Molecular Weight (g/mol) 444.24
MDL Number MFCD00043297
InChI Key YWMRPVUMBTVUEX-UHFFFAOYSA-N
Synonym tetrakis acetonitrile palladium ii tetrafluoroborate, acetonitrile; palladium 2+ ; ditetrafluoroborate, pd mecn 4 bf4 2, tetrakis acetonitrile palladium ii, palladium ii tetrafluoroborate tetraacetonitrile complex
PubChem CID 2734560
SMILES [Pd++].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Melting Point 230°C (decomposition)
Quantity 0.25 g
UN Number UN2811
Sensitivity Air Sensitive
Solubility Information Soluble in water
IUPAC Name palladium(2+) tetrakis(acetonitrile) bis(tetrafluoroboranuide)
Formula Weight 444.22
Concentration or Composition (by Analyte or Components) Pd 23.5%
Physical Form Powder
Chemical Name or Material Tetrakis(acetonitrile)palladium(II) tetrafluoroborate
Hazard Category H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P271-P304+P340-P312
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.