Learn More
Tetrakis(trimethylsilyl)silane, 98%
Description
It is used as a starting material for the synthesis of tris(trimethylsilyl)silyl lithium with methyl lithium and in the preparation of lithium bis[(TMS)3silyl]cuprate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as a starting material for the synthesis of tris(trimethylsilyl)silyl lithium with methyl lithium and in the preparation of lithium bis[(TMS)3silyl]cuprate.
Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
Specifications
Specifications
| Melting Point | 266°C to 268°C (sublimation) |
| Quantity | 1 g |
| Formula Weight | 320.85 |
| Percent Purity | 98% |
| Chemical Name or Material | Tetrakis(trimethylsilyl)silane |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.