Alfa Aesar™ Tetrakisepoxy cyclosiloxane

Manufacturer: Alfa Aesar™ 4696014

Quantity 25g

 View more versions of this product

Catalog No. AA4696014

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 257284-60-9
    Molecular Formula C28H56O12Si4
    Formula Weight 697.09
    MDL Number MFCD22380706
    Synonym 1,3,5,7-Tetramethyl-1,3,5,7-tetrakis[3-(glycidyloxy)propyl]cyclotetrasiloxane
    Chemical Name or Material Tetrakisepoxy cyclosiloxane

    Specifications

    Formula Weight 697.09
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Liquid
    Molecular Formula C28H56O12Si4
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2°C to 8°C
    Quantity 25g
    MDL Number MFCD22380706
    CAS 257284-60-9
    Synonym 1,3,5,7-Tetramethyl-1,3,5,7-tetrakis[3-(glycidyloxy)propyl]cyclotetrasiloxane
    Health Hazard 1 Warning
    Color Colorless
    Chemical Name or Material Tetrakisepoxy cyclosiloxane

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit