Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethyl orthocarbonate, 95%

Catalog No. p-7057192
Click to view available options
Quantity:
1 g
5 g

CAS: 1850-14-2 | C5H12O4 | 136.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1850-14-2
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.147
MDL Number MFCD00008473
InChI Key AHJWSRRHTXRLAQ-UHFFFAOYSA-N
Synonym tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc
PubChem CID 74613
IUPAC Name tetramethoxymethane
SMILES COC(OC)(OC)OC

Specifications

Melting Point -5°C
Color Colorless
Density 1.017 g/mL
Boiling Point 114°C
Flash Point 22°C (72°F)
Refractive Index 1.385
Linear Formula C(OCH3)4
Quantity 1 g
UN Number UN3272
Beilstein 1737682
Sensitivity Moisture sensitive
Formula Weight 136.15
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tetramethyl orthocarbonate
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 217-438-5
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.