Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethyl orthocarbonate, 99%

Catalog No. ac158110050
Click to view available options
Quantity:
5 g
25 g

CAS: 1850-14-2 | C5H12O4 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1850-14-2
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.15
InChI Key AHJWSRRHTXRLAQ-UHFFFAOYSA-N
Synonym tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc
PubChem CID 74613
IUPAC Name tetramethoxymethane
SMILES COC(OC)(OC)OC

Specifications

Melting Point -5.0°C
Density 1.0200g/mL
Boiling Point 114.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Refractive Index 1.3845
Linear Formula C(OCH3)4
Quantity 5 g
Beilstein 03,II,4
Fieser 03,285
Specific Gravity 1.02
Formula Weight 136.15
Percent Purity 99%
Chemical Name or Material Tetramethyl orthocarbonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 217-438-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.