Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEV Cleavage Site Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB397068 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397068 25 μg
NBP237831 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB397068 Supplier Novus Biologicals Supplier No. NBP23783125UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TEV Cleavage Site Polyclonal specifically detects TEV Cleavage Site in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TEV Cleavage Site
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 μg/mL
Formulation BBS (pH 8.4) with No Preservative
Host Species Rabbit
Immunogen Peptide representing the cleavage site recognized by TEV enzyme
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product has been successfully used in Western blot applications. The immunogen is a peptide representing the cleavage site recognized by TEV enzyme.
Target Species Non-species specific
Content And Storage Store at 4°C. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.