Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEV Cleavage Site Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$289.00 - $499.50
Specifications
Antigen | TEV Cleavage Site |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
TEV Cleavage Site Polyclonal specifically detects TEV Cleavage Site in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TEV Cleavage Site | |
Western Blot | |
Unconjugated | |
RUO | |
BBS (pH 8.4) with No Preservative | |
IgG | |
Affinity Purified |
0.5 mg/mL | |
Polyclonal | |
Rabbit | |
Non-species specific | |
Peptide representing the cleavage site recognized by TEV enzyme | |
Primary | |
This product has been successfully used in Western blot applications. The immunogen is a peptide representing the cleavage site recognized by TEV enzyme. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title