Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TEV Protease Antibody (9A10-4C3), Novus Biologicals™
SDP

Catalog No. NB413895 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 ug
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB413895 25 ug
NBP229909 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB413895 Supplier Novus Biologicals Supplier No. NBP22990925UG

Mouse Monoclonal Antibody

TEV Protease Monoclonal specifically detects TEV Protease in Non-species specific samples. It is validated for Western Blot.

Specifications

Antigen TEV Protease
Applications Western Blot
Classification Monoclonal
Clone 9A10-4C3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2000
Gene Accession No. Q0GDU8
Gene Alias P1 Protease, Tobacco Etch Virus Protease
Host Species Mouse
Immunogen KLH conjugate peptide representing the cleavage site recognized by TEV enzyme
Purification Method Protein A purified
Quantity 25 ug
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1502321
Test Specificity TAG.
Target Species Non-species specific
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.