Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TEV Protease Antibody (9A10-4C3), Novus Biologicals™

Mouse Monoclonal Antibody
$499.50
Specifications
| Antigen | TEV Protease |
|---|---|
| Clone | 9A10-4C3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 - 1:2000 |
| Applications | Western Blot |
Description
TEV Protease Monoclonal specifically detects TEV Protease in Non-species specific samples. It is validated for Western Blot.Specifications
| TEV Protease | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Non-species specific | |
| P1 Protease, Tobacco Etch Virus Protease | |
| KLH conjugate peptide representing the cleavage site recognized by TEV enzyme | |
| Primary | |
| Store at 4C. Do not freeze. |
| 9A10-4C3 | |
| Western Blot 1:1000 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q0GDU8 | |
| 1502321 | |
| IgG1 | |
| Protein A purified | |
| TAG. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title