Learn More
Description
Specifications
Specifications
| Antigen | TEX101 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | cancer/testis antigen 131, Cell surface receptor NYD-SP8, CT131, MGC4766, NYD-SP8, PRO1884, Scleroderma-associated autoantigen, SGRGGTPR867, Spermatogenesis-related gene protein, TES101RP, testis expressed 101, testis expressed sequence 101, testis-expressed protein 101, testis-specific protein TES101RP |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of Human TEX101 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
