Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tex14 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220605 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220605 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220605 Supplier Novus Biologicals Supplier No. NBP220605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tex14 Polyclonal specifically detects Tex14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Tex14
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias cancer/testis antigen 113, CT113, Protein kinase-like protein SgK307, SGK307, Sugen kinase 307, testis expressed 14, testis expressed sequence 14, testis-expressed protein 14, Testis-expressed sequence 14
Gene Symbols TEX14
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human TEX14. The exact sequence is proprietary.
Molecular Weight of Antigen 168 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56155
Test Specificity Human. Species reactivity based on sequence identity: Mouse (85%), Bovine (89%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.