Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tex14 Antibody, Novus Biologicals™
SDP

Catalog No. NBP154999 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP154999 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP154999 Supplier Novus Biologicals Supplier No. NBP154999
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tex14 Polyclonal specifically detects Tex14 in Human samples. It is validated for Western Blot.

Specifications

Antigen Tex14
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8IWB6
Gene Alias cancer/testis antigen 113, CT113, Protein kinase-like protein SgK307, SGK307, Sugen kinase 307, testis expressed 14, testis expressed sequence 14, testis-expressed protein 14, Testis-expressed sequence 14
Gene Symbols TEX14
Host Species Rabbit
Immunogen Synthetic peptides corresponding to TEX14(testis expressed 14) The peptide sequence was selected from the C terminal of TEX14. Peptide sequence ASSDTLVAVEKSYSTSSPIEEDFEGIQGAFAQPQVSGEEKFQMRKILGKN.
Molecular Weight of Antigen 160 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56155
Test Specificity Rat: 79%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Rat, Equine, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.