Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFAP4 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291530 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291530 Supplier Novus Biologicals Supplier No. NBP146201F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TFAP4 Polyclonal antibody specifically detects TFAP4 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen TFAP4
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias Activating enhancer-binding protein 4, AP-4, BHLHC41, bHLHc41Class C basic helix-loop-helix protein 41, transcription factor AP-4, transcription factor AP-4 (activating enhancer binding protein 4), transcription factor AP-4 (activating enhancer-binding protein 4)
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 50 and 100 of human Activating Enhancer-Binding Protein 4 using the numbering given in entry NP_003214.1 (GeneID 7023).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7023
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.