Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFAP4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146201 Supplier Novus Biologicals Supplier No. NBP146201
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TFAP4 Polyclonal specifically detects TFAP4 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen TFAP4
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Alias Activating enhancer-binding protein 4, AP-4, BHLHC41, bHLHc41Class C basic helix-loop-helix protein 41, transcription factor AP-4, transcription factor AP-4 (activating enhancer binding protein 4), transcription factor AP-4 (activating enhancer-binding protein 4)
Gene Symbols TFAP4
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 50 and 100 of human Activating Enhancer-Binding Protein 4 using the numbering given in entry NP_003214.1 (GeneID 7023).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7023
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.