Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFB1M Antibody (OTI6G4), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N274490A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274490A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274490A700 Supplier Novus Biologicals Supplier No. NBP274490AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TFB1M Monoclonal specifically detects TFB1M in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen TFB1M
Applications Western Blot
Classification Monoclonal
Clone OTI6G4
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CGI75, CGI-75, dimethyladenosine transferase 1, mitochondrial, EC 2.1.1, EC 2.1.1.-, h-mtTFB, h-mtTFB1, homolog of yeast mitochondrial transcription factor B, hTFB1M, Mitochondrial 12S rRNA dimethylase 1, mitochondrial dimethyladenosine transferase 1, Mitochondrial transcription factor B1, mtTFB, mtTFB1, S-adenosylmethionine-6-N′, N′-adenosyl(rRNA) dimethyltransferase 1, transcription factor B1, mitochondrial
Gene Symbols TFB1M
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 116-346 of human TFB1M (NP_057104) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51106.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.