Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFCP2L1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB184030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184030 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184030 Supplier Novus Biologicals Supplier No. NBP282042H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TFCP2L1 Polyclonal antibody specifically detects TFCP2L1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen TFCP2L1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias CRTR1CRTR-1, LBP-9, LBP9CP2-related transcriptional repressor 1, transcription factor CP2-like 1, transcription factor CP2-like protein 1, Transcription factor LBP-9
Host Species Rabbit
Immunogen TFCP2L1 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human TFCP2L1. The immunogen is located within the first 50 amino acids of TFCP2L1. Amino Acid Squence: HTQPEHYNQHNSGSY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 29842
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.