Learn More
Description
Specifications
Specifications
| Antigen | TFEB |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 350 |
| Dilution | Western Blot |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB |
| Gene Symbols | TFEB |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 325 and 375 of human Transcription Factor EB using the numbering given in entry NP_009093.1 (GeneID 7942). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
