Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFEB Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP212758G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212758G 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212758G Supplier Novus Biologicals Supplier No. NBP212758G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TFEB Polyclonal specifically detects TFEB in Human samples. It is validated for Western Blot.

Specifications

Antigen TFEB
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB
Gene Symbols TFEB
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 325 and 375 of human Transcription Factor EB using the numbering given in entry NP_009093.1 (GeneID 7942).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7942
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.