Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TFEB Antibody, Novus Biologicals™
SDP

Catalog No. NB407591 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB407591 20 μg
NB10093447 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB407591 Supplier Novus Biologicals Supplier No. NB1009344720UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

TFEB Polyclonal specifically detects TFEB in Human, Rat samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen TFEB
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 μg/mL, Peptide ELISA Detection limit 1:1000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P19484
Gene Alias ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB
Gene Symbols TFEB
Host Species Goat
Immunogen Peptide with sequence C-QKDLQKSRELENH corresponding to internal region according to NP_009093.1.
Purification Method Affinity Purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7942
Test Specificity This antibody is expected not to cross-react to TFE3 and MITF.
Target Species Human, Rat, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.