Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TFEB Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
$231.00 - $487.50
Specifications
Antigen | TFEB |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10093447
![]() |
Novus Biologicals
NB10093447 |
0.1 mg |
Each for $487.50
|
|
|||||
NB407591
![]() |
Novus Biologicals
NB1009344720UG |
20 μg |
Each for $231.00
|
|
|||||
Description
TFEB Polyclonal specifically detects TFEB in Human, Rat samples. It is validated for Western Blot, Peptide ELISA.Specifications
TFEB | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
ALPHATFEB, BHLHE35, Class E basic helix-loop-helix protein 35, T-cell transcription factor EB, TCFEBbHLHe35, transcription factor EB | |
TFEB | |
IgG | |
Affinity Purified |
0.5 mg/mL | |
Polyclonal | |
Goat | |
Human, Rat | |
P19484 | |
7942 | |
Peptide with sequence C-QKDLQKSRELENH corresponding to internal region according to NP_009093.1. | |
Primary | |
This antibody is expected not to cross-react to TFE3 and MITF. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title